This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03355351_m1
See other KIAA1524 GE Assays ›
Gene Symbol:
KIAA1524
Gene Name:
KIAA1524
Gene Aliases:
-
Chromosome Location:
Chr.1: 88827764 - 88854919 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03355351_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KIAA1524
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030785.2 NP_001025956.1 17 - 18 2312 167
XM_025149325.1 XP_025005093.1 16 - 17 2217 167
XM_025149326.1 XP_025005094.1 16 - 17 2039 167
GenBank mRNA AJ719387.1 - 17 - 18 2313 167

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0