Assay Details
Gene Symbol: ZDHHC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031438.1 | NP_001026609.1 | 4 - 5 | 580 | 84 | |
XM_015280103.2 | XP_015135589.1 | 4 - 5 | 478 | 84 | ||
XM_015280105.2 | XP_015135591.1 | 3 - 4 | 772 | 84 | ||
XM_015280106.2 | XP_015135592.1 | 4 - 5 | 590 | 84 | ||
XM_025144727.1 | XP_025000495.1 | 4 - 5 | 590 | 84 | ||
GenBank mRNA | AJ851604.1 | - | 4 - 5 | 580 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
