This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03352094_m1
See other KLHL9 GE Assays ›
Gene Symbol:
KLHL9
Gene Name:
kelch like family member 9
Gene Aliases:
KLHL13
Chromosome Location:
Chr.4: 3197426 - 3265739 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03352094_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KLHL9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031116.1 NP_001026287.1 4 - 5 341 142
XM_015278239.2 XP_015133725.1 3 - 4 294 142
XM_015278240.2 XP_015133726.1 2 - 3 392 142
XM_015278241.2 XP_015133727.1 2 - 3 534 142
XM_015278242.2 XP_015133728.1 3 - 4 291 142
XM_025149555.1 XP_025005323.1 4 - 5 321 142
GenBank mRNA AJ719801.1 - 4 - 5 341 142

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
protein ubiquitination
regulation of cytokinesis
cell division
ubiquitin-protein transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0