This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03351090_m1
See other SRSF6 GE Assays ›
Gene Symbol:
SRSF6
Gene Name:
serine and arginine rich splicing factor 6
Gene Aliases:
SFRS6
Chromosome Location:
Chr.20: 119215 - 123854 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03351090_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRSF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030843.2 NP_001026014.1 4 - 5 773 81
GenBank mRNA AJ720575.1 - 4 - 5 717 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm