This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03348083_m1
See other CALM2 GE Assays ›
Gene Symbol:
CALM2
Gene Name:
calmodulin 2
Gene Aliases:
CALM
Chromosome Location:
Chr.3: 27251680 - 27263186 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03348083_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CALM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205005.1 NP_990336.1 4 - 5 341 135
GenBank mRNA AF081672.1 - 4 - 5 341 135
AJ720728.1 - 4 - 5 135
M36167.1 - 3 - 4 282 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
calcium-mediated signaling
regulation of catalytic activity
calcium ion binding
protein C-terminus binding
myosin binding
enzyme regulator activity
CH domain binding
disordered domain specific binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header