This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03346955_m1
See other PDGFB GE Assays ›
Gene Symbol:
PDGFB
Gene Name:
platelet derived growth factor subunit B
Gene Aliases:
-
Chromosome Location:
Chr.1: 50720076 - 50737739 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03346955_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDGFB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204270.4 NP_989601.2 1 - 2 198 89
XM_015284369.2 XP_015139855.2 1 - 2 1074 89
XM_015284373.2 XP_015139859.2 2 - 3 1050 89
XM_015284377.2 XP_015139863.2 2 - 3 953 89
XM_025150351.1 XP_025006119.1 2 - 3 980 89
GenBank mRNA AB031025.1 - 1 - 2 159 89
BU372893.1 - 1 - 2 198 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0