This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03346228_m1
See other RACK1 GE Assays ›
Gene Symbol:
RACK1
Gene Name:
receptor for activated C kinase 1
Gene Aliases:
C12-3, GNB2L1, c12.3
Chromosome Location:
Chr.16: 2526354 - 2529817 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03346228_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RACK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004378.2 NP_001004378.1 1 - 2 216 64
GenBank mRNA AJ392770.1 - 1 - 2 216 64
AY393848.1 - 1 - 2 114 64
CR386189.1 - 1 - 2 196 64
M24193.1 - 1 - 2 196 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
negative regulation of gene expression
protein ubiquitination
negative regulation of Wnt signaling pathway
negative regulation of cell growth
positive regulation of cell migration
positive regulation of protein complex assembly
negative regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of protein localization
negative regulation of peptidyl-serine phosphorylation
positive regulation of Golgi to plasma membrane protein transport
pigmentation
positive regulation of GTPase activity
negative regulation of phagocytosis
regulation of cell division
positive regulation of cyclic-nucleotide phosphodiesterase activity
regulation of cell cycle
negative regulation of protein kinase B signaling
positive regulation of mitochondrial depolarization
negative regulation of protein tyrosine kinase activity
cellular response to glucose stimulus
cellular response to growth factor stimulus
rescue of stalled ribosome
negative regulation of hydrogen peroxide-induced neuron death
regulation of establishment of cell polarity
positive regulation of gastrulation
positive regulation of intrinsic apoptotic signaling pathway
protein kinase C binding
ion channel inhibitor activity
cysteine-type endopeptidase activator activity involved in apoptotic process
enzyme binding
protein phosphatase binding
protein tyrosine kinase inhibitor activity
cyclin binding
receptor tyrosine kinase binding
signaling adaptor activity
SH2 domain binding
protein homodimerization activity
ribosome binding
BH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0