This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03345725_m1
See other GART GE Assays ›
Gene Symbol:
GART
Gene Name:
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
Gene Aliases:
AIRS, GARS, GART-A
Chromosome Location:
Chr.1: 106695137 - 106735453 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03345725_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GART
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001469.1 NP_001001469.1 2 - 3 197 69
XM_015299217.2 XP_015154703.1 4 - 5 406 69
XM_015299227.1 XP_015154713.1 5 - 6 519 69
XM_015299230.2 XP_015154716.1 4 - 5 459 69
XM_015299235.2 XP_015154721.1 3 - 4 362 69
XM_025153908.1 XP_025009676.1 4 - 5 489 69
XM_025154010.1 XP_025009778.1 3 - 4 362 69
GenBank mRNA CR523902.1 - 3 - 4 342 69
FJ977572.1 - 1 - 2 157 69
X54200.1 - 197 69
X56339.1 - 197 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide biosynthetic process
'de novo' IMP biosynthetic process
adenine biosynthetic process
phosphoribosylamine-glycine ligase activity
phosphoribosylformylglycinamidine cyclo-ligase activity
phosphoribosylglycinamide formyltransferase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0