This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03344087_m1
See other CAMP GE Assays ›
Gene Symbol:
CAMP
Gene Name:
cathelicidin antimicrobial peptide
Gene Aliases:
CATH-2, CATH2, CATHL2, CMAP27
Chromosome Location:
Chr.2: 4074457 - 4076222 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03344087_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAMP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024830.2 NP_001020001.2 2 - 3 274 101
GenBank mRNA AY817057.1 - 2 - 3 274 101
DQ092352.1 - 2 - 3 274 101
FJ938358.1 - 2 - 3 274 101
HQ640432.1 - 3 - 4 274 101
KT962965.1 - 3 - 4 274 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s) Process(es)

cell death
negative regulation of gene expression
cytolysis
innate immune response
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
antimicrobial humoral immune response mediated by antimicrobial peptide
lipopolysaccharide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0