This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03341158_m1
See other PDHA2 GE Assays ›
Gene Symbol:
PDHA2
Gene Name:
pyruvate dehydrogenase (lipoamide) alpha 2
Gene Aliases:
PDHA1
Chromosome Location:
Chr.1: 121691902 - 121703921 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03341158_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDHA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012544.2 NP_001012562.1 9 - 10 972 89
XM_015302559.2 XP_015158045.1 10 - 11 1013 89
GenBank mRNA AJ851474.1 - 9 - 10 973 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0