This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03339710_m1
See other IGFBP2 GE Assays ›
Gene Symbol:
IGFBP2
Gene Name:
insulin like growth factor binding protein 2
Gene Aliases:
IGFBP-2
Chromosome Location:
Chr.7: 23252168 - 23328028 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03339710_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGFBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205359.1 NP_990690.1 2 - 3 657 82
XM_015289789.2 XP_015145275.1 2 - 3 342 82
XM_015289790.2 XP_015145276.1 2 - 3 315 82
XM_015289791.2 XP_015145277.1 2 - 3 247 82
XM_025152151.1 XP_025007919.1 3 - 4 375 82
XM_025152152.1 XP_025007920.1 2 - 3 351 82
XM_025152153.1 XP_025007921.1 2 - 3 273 82
GenBank mRNA AJ544105.1 - 2 - 3 321 82
U15086.1 - 2 - 3 657 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

regulation of growth
regulation of insulin-like growth factor receptor signaling pathway
insulin-like growth factor I binding
insulin-like growth factor II binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0