This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338898_m1
See other HPRT1 GE Assays ›
Gene Symbol:
HPRT1
Gene Name:
hypoxanthine phosphoribosyltransferase 1
Gene Aliases:
HPRT
Chromosome Location:
Chr.4: 4025994 - 4041861 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338898_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HPRT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204848.1 NP_990179.1 6 - 7 538 72
GenBank mRNA AJ132697.1 - 6 - 7 538 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Endogenous Controls Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide biosynthetic process
purine ribonucleoside salvage
adenine salvage
guanine salvage
GMP salvage
IMP salvage
hypoxanthine salvage
positive regulation of dopamine metabolic process
GMP catabolic process
IMP metabolic process
hypoxanthine metabolic process
nucleotide binding
magnesium ion binding
guanine binding
purine nucleobase binding
hypoxanthine phosphoribosyltransferase activity
identical protein binding
guanine phosphoribosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0