This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338882_g1
See other RAP1GAP3 GE Assays ›
Gene Symbol:
RAP1GAP3
Gene Name:
RAP1 GTPase activating protein 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 78362968 - 78418997 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338882_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAP1GAP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204843.3 NP_990174.1 9 - 10 701 122
XM_015293174.2 XP_015148660.1 9 - 10 749 122
XM_015293180.2 XP_015148666.1 9 - 10 749 122
XM_025142814.1 XP_024998582.1 10 - 11 834 122
XM_025142833.1 XP_024998601.1 9 - 10 798 122
XM_025142865.1 XP_024998633.1 10 - 11 834 122
XM_025142985.1 XP_024998753.1 10 - 11 834 122
XM_025143003.1 XP_024998771.1 10 - 11 834 122
XM_025143026.1 XP_024998794.1 9 - 10 743 122
XM_025143049.1 XP_024998817.1 9 - 10 648 122
XM_025143108.1 XP_024998876.1 9 - 10 749 122
XM_025143121.1 XP_024998889.1 9 - 10 742 122
XM_025143148.1 XP_024998916.1 5 - 6 1056 122
GenBank mRNA AF151966.2 - 9 - 10 701 122
CR386855.1 - 6 - 7 984 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0