This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03338881_m1
See other RAP1GAP3 GE Assays ›
Gene Symbol:
RAP1GAP3
Gene Name:
RAP1 GTPase activating protein 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 78362968 - 78418997 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03338881_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAP1GAP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204843.3 NP_990174.1 8 - 9 642 96
XM_015293174.2 XP_015148660.1 8 - 9 690 96
XM_015293180.2 XP_015148666.1 8 - 9 690 96
XM_025142814.1 XP_024998582.1 9 - 10 775 96
XM_025142833.1 XP_024998601.1 8 - 9 739 96
XM_025142865.1 XP_024998633.1 9 - 10 775 96
XM_025142985.1 XP_024998753.1 9 - 10 775 96
XM_025143003.1 XP_024998771.1 9 - 10 775 96
XM_025143026.1 XP_024998794.1 8 - 9 684 96
XM_025143049.1 XP_024998817.1 8 - 9 589 96
XM_025143108.1 XP_024998876.1 8 - 9 690 96
XM_025143121.1 XP_024998889.1 8 - 9 683 96
XM_025143148.1 XP_024998916.1 4 - 5 997 96
GenBank mRNA AF151966.2 - 8 - 9 642 96
CR386855.1 - 5 - 6 925 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0