This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03337980_m1
See other IL6 GE Assays ›
Gene Symbol:
IL6
Gene Name:
interleukin 6
Gene Aliases:
CHIL-6, IL-6, interleukin-6
Chromosome Location:
Chr.2: 30862822 - 30873241 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03337980_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204628.1 NP_989959.1 3 - 4 559 61
XM_015281283.2 XP_015136769.1 2 - 3 1126 61
GenBank mRNA AB302327.1 - 3 - 4 396 61
AJ309540.1 - 3 - 4 559 61
HM179640.1 - 3 - 4 550 61
JQ897539.1 - 3 - 4 537 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

acute-phase response
vascular endothelial growth factor production
response to activity
glucose homeostasis
regulation of insulin secretion
regulation of glucagon secretion
interleukin-6-mediated signaling pathway
cellular response to cold
T-helper 17 cell lineage commitment
hepatocyte proliferation
liver regeneration
positive regulation of STAT cascade
cytokine activity
interleukin-6 receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0