This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03337637_m1
See other VEGFD GE Assays ›
Gene Symbol:
VEGFD
Gene Name:
vascular endothelial growth factor D
Gene Aliases:
FIGF
Chromosome Location:
Chr.1: 123415537 - 123447158 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03337637_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VEGFD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204568.2 NP_989899.2 4 - 5 1092 81
XM_015273093.2 XP_015128579.1 4 - 5 7040 81
GenBank mRNA AF479650.1 - 4 - 5 659 81
EF569672.1 - 4 - 5 639 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0