This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03332550_m1
See other FAM213A GE Assays ›
Gene Symbol:
FAM213A
Gene Name:
family with sequence similarity 213 member A
Gene Aliases:
C10ORF58, C6H10orf58, PAMM, PRXL2A, SELENOU
Chromosome Location:
Chr.6: 5107184 - 5118458 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03332550_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM213A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193518.1 NP_001180447.1 2 - 3 314 130
NM_001193519.2 NP_001180448.1 2 - 3 305 130
GenBank mRNA AJ720950.1 - 2 - 3 306 130
BM491988.1 - 2 - 3 298 130
BU119844.1 - 2 - 3 210 130
BU422656.1 - 2 - 3 209 130
KP278239.1 - 1 - 2 184 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
cellular oxidant detoxification
antioxidant activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0