Assay Details
Gene Symbol: CHIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420705.6 | XP_420705.1 | 1 - 2 | 322 | 126 | |
GenBank mRNA | CR353754.1 | - | 1 - 2 | 177 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
