This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03328627_m1
See other HSD17B7 GE Assays ›
Gene Symbol:
HSD17B7
Gene Name:
hydroxysteroid 17-beta dehydrogenase 7
Gene Aliases:
-
Chromosome Location:
Chr.8: 4187219 - 4193089 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03328627_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD17B7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277506.1 NP_001264435.1 7 - 8 862 75
NM_001277507.1 NP_001264436.1 6 - 7 769 75
GenBank mRNA BU265534.1 - 713 75
BX933486.2 - 7 - 8 816 75
BX935618.1 - 7 - 8 818 75
BX950661.1 - 6 - 7 755 75
CR352574.1 - 7 - 8 792 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0