This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03321251_m1
See other DUPD1 GE Assays ›
Gene Symbol:
DUPD1
Gene Name:
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 15803539 - 15823046 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03321251_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUPD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168006.1 NP_001161478.1 2 - 3 255 66
GenBank mRNA BX935843.1 - 2 - 3 261 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0