This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03321108_m1
See other PCMT1L GE Assays ›
Gene Symbol:
PCMT1L
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase-like
Gene Aliases:
-
Chromosome Location:
Chr.5: 1132951 - 1136917 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03321108_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMT1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277402.1 NP_001264331.1 5 - 6 474 127
NM_001277404.1 NP_001264333.1 5 - 6 474 127
GenBank mRNA BU383489.1 - 2 - 3 132 128
BX929703.2 - 5 - 6 459 127
BX935325.1 - 5 - 6 437 127
BX950403.1 - 5 - 6 437 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0