Assay Details
Gene Symbol: IER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640667.1 | XP_023496435.1 | 523 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: IER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640667.1 | XP_023496435.1 | 523 | 75 |
Set Membership: |
![]() ![]() ![]() ![]() |