This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07067447_g1
See other STING1 GE Assays ›
Gene Symbol:
STING1
Gene Name:
stimulator of interferon response cGAMP interactor 1
Gene Aliases:
TMEM173
Chromosome Location:
Chr.14: 36509832 - 36515340 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07067447_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STING1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005599365.3 XP_005599422.1 977 62
XM_005599366.3 XP_005599423.1 962 62
XM_023617601.1 XP_023473369.1 1021 62
XM_023617602.1 XP_023473370.1 1013 62
XM_023617603.1 XP_023473371.1 1227 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header