This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07054931_m1
See other GJC1 GE Assays ›
Gene Symbol:
GJC1
Gene Name:
gap junction protein gamma 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 18879560 - 18925502 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07054931_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001489028.4 XP_001489078.1 375 124

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001489028.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Equus caballus

Panther Classification:

Molecular Function -

gap junction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0