This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07038784_m1
See other GPR34 GE Assays ›
Gene Symbol:
GPR34
Gene Name:
G protein-coupled receptor 34
Gene Aliases:
-
Chromosome Location:
Chr.X: 35107773 - 35114878 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07038784_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014728818.2 XP_014584304.1 176 85
XM_014728819.2 XP_014584305.1 210 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014728818.2, XM_014728819.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Equus caballus

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0