Assay Details
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642990.1 | XP_023498758.1 | 366 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642990.1 | XP_023498758.1 | 366 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() |