Assay Details
| Gene Symbol: FITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918360.5 | XP_001918395.3 | 2804 | 111 | ||
More Information
Set Membership: |
|

FITM1| Gene Symbol: FITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918360.5 | XP_001918395.3 | 2804 | 111 | ||
Set Membership: |
|