This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06968321_m1
See other LOC100063291 GE Assays ›
Gene Symbol:
LOC100063291
Gene Name:
N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase
Gene Aliases:
GCNT2
Chromosome Location:
Chr.20: 10944505 - 11030995 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06968321_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100063291
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023624445.1 XP_023480213.1 111 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_023624445.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Equus caballus

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0