This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06961188_m1
See other CFAP91 GE Assays ›
Gene Symbol:
CFAP91
Gene Name:
cilia and flagella associated protein 91
Gene Aliases:
MAATS1
Chromosome Location:
Chr.19: 41642579 - 41704265 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06961188_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CFAP91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014732868.2 XP_014588354.2 2223 72
XM_023623702.1 XP_023479470.1 2676 72
XM_023623703.1 XP_023479471.1 2676 72
XM_023623705.1 XP_023479473.1 2636 72
XM_023623707.1 XP_023479475.1 1858 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header