This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06956204_m1
See other SEPTIN2 GE Assays ›
Gene Symbol:
SEPTIN2
Gene Name:
septin 2
Gene Aliases:
SEPT2
Chromosome Location:
Chr.6: 26623830 - 26658475 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06956204_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEPTIN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023644074.1 XP_023499842.1 326 75
XM_023644075.1 XP_023499843.1 480 75
XM_023644076.1 XP_023499844.1 267 75
XM_023644077.1 XP_023499845.1 408 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Equus caballus

Panther Classification:

Molecular Function -

cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0