This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06954555_g1
See other RPL15 GE Assays ›
Gene Symbol:
RPL15
Gene Name:
ribosomal protein L15
Gene Aliases:
-
Chromosome Location:
Chr.16: 60573975 - 60578194 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06954555_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001494613.5 XP_001494663.1 128 91

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001494613.5 transcripts.

Set Membership:

Amplicon length between 86 and 100 Equus caballus

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0