This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06951116_m1
See other CYRIA GE Assays ›
Gene Symbol:
CYRIA
Gene Name:
CYFIP related Rac1 interactor A
Gene Aliases:
FAM49A
Chromosome Location:
Chr.15: 79252368 - 79355190 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06951116_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYRIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005600202.3 XP_005600259.1 2037 136
XM_005600208.3 XP_005600265.1 4225 136
XM_014731299.2 XP_014586785.1 1130 136
XM_023619492.1 XP_023475260.1 1141 136
XM_023619493.1 XP_023475261.1 1926 136
XM_023619494.1 XP_023475262.1 2065 136
XM_023619496.1 XP_023475264.1 1906 136
XM_023619497.1 XP_023475265.1 1661 136
XM_023619498.1 XP_023475266.1 1515 136
XM_023619499.1 XP_023475267.1 1285 136
XM_023619500.1 XP_023475268.1 1287 136

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0