This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06948690_m1
See other VAMP5 GE Assays ›
Gene Symbol:
VAMP5
Gene Name:
vesicle associated membrane protein 5
Gene Aliases:
-
Chromosome Location:
Chr.15: 19574482 - 19581314 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06948690_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAMP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001498842.4 XP_001498892.1 153 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001498842.4 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0