This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04952226_m1
See other MMUT GE Assays ›
Gene Symbol:
MMUT
Gene Name:
methylmalonyl-CoA mutase
Gene Aliases:
MUT
Chromosome Location:
Chr.20: 48459519 - 48490356 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04952226_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MMUT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001309313.2 NP_001296242.1 5 - 6 1369 117
XM_001502799.5 XP_001502849.1 1467 117
XM_014734210.2 XP_014589696.1 1512 117
XM_014734211.2 XP_014589697.1 1476 117
GenBank mRNA JL640521.1 - 2 - 3 272 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Equus caballus Probe spans exons

Panther Classification:

Molecular Function -

mutase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0