Assay Details
Gene Symbol: MIR9078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128101.1 | 1 | 123 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear receptor binding SET domain protein 2 | NSD2 | WHSC1 | -53.4kb | Anti-sense | Chr. 3 - 120084061 - 120172945 on NCBI Build EquCab3.0 |
Downstream | fibroblast growth factor receptor 3 | FGFR3 | 4.6kb | Anti-sense | Chr. 3 - 120231097 - 120247930 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
