This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04648114_s1
See other MIR8997 GE Assays ›
Gene Symbol:
MIR8997
Gene Name:
microRNA mir-8997
Gene Aliases:
eca-mir-8997
Chromosome Location:
Chr.7: 46046577 - 46046715 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04648114_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR8997
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_128005.1 1 25 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adhesion G protein-coupled receptor L1 ADGRL1 LPHN1 -18.7kb Sense Chr. 7 - 45987073 - 46027887 on NCBI Build EquCab3.0
Downstream protein kinase cAMP-activated catalytic subunit alpha PRKACA 2.0kb Sense Chr. 7 - 46048702 - 46065128 on NCBI Build EquCab3.0

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Equus caballus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0