This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04647953_s1
See other MIR342 GE Assays ›
Gene Symbol:
MIR342
Gene Name:
microRNA mir-342
Gene Aliases:
eca-mir-342
Chromosome Location:
Chr.24: 42491171 - 42491269 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04647953_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR342
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033012.1 1 51 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA mir-8954 MIR8954 eca-mir-8954 -121.9kb Anti-sense Chr. 24 - 42369100 - 42369242 on NCBI Build EquCab3.0
Downstream delta 4-desaturase,
sphingolipid 2
DEGS2 41.5kb Anti-sense Chr. 24 - 42532738 - 42550975 on NCBI Build EquCab3.0

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Equus caballus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0