Assay Details
Gene Symbol: MIR491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032995.1 | 1 | 39 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | focadhesin | FOCAD | Sense | Chr. 23 - 39349869 - 39619804 on NCBI Build EquCab3.0 | ||
Downstream | 3-hydroxyacyl-CoA dehydratase 4 | HACD4 | PTPLAD2 | 244.8kb | Anti-sense | Chr. 23 - 39623439 - 39644444 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
