This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04320704_m1
See other SELENOF GE Assays ›
Gene Symbol:
SELENOF
Gene Name:
selenoprotein F
Gene Aliases:
SEP15
Chromosome Location:
Chr.5: 74014800 - 74051683 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04320704_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SELENOF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170338.1 NP_001163809.1 4 - 5 637 95
GenBank mRNA CD464773.1 - 4 - 5 373 95
CD536845.1 - 4 - 5 362 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Equus caballus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0