This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04319849_s1
See other CCR5 GE Assays ›
Gene Symbol:
CCR5
Gene Name:
C-C motif chemokine receptor 5
Gene Aliases:
-
Chromosome Location:
Chr.16: 41954183 - 41980124 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04319849_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCR5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001091534.1 NP_001085003.1 1 1021 65
XM_023619764.1 XP_023475532.1 1085 65
XM_023619765.1 XP_023475533.1 1192 65
GenBank mRNA DQ629590.1 - 2 1317 65
JL619065.1 - 2 1079 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0