This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03445589_s1
See other DAO.1 GE Assays ›
Gene Symbol:
dao.1
Gene Name:
D-amino-acid oxidase, tandem duplicate 1
Gene Aliases:
wu:fb55b11, zgc:113922
Chromosome Location:
Chr.5: 20187877 - 20195419 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03445589_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dao.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033740.1 NP_001028912.1 11 1146 143
XM_005165183.3 XP_005165240.1 11 1191 144
GenBank mRNA BC097016.1 - 11 1146 143

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

proline catabolic process
D-amino acid catabolic process
D-serine catabolic process
D-amino acid metabolic process
oxidation-reduction process
D-alanine catabolic process
D-amino-acid oxidase activity
oxidoreductase activity
FAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0