This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03441109_m1
See other EEF1A1L2 GE Assays ›
Gene Symbol:
eef1a1l2
Gene Name:
eukaryotic translation elongation factor 1 alpha 1, like 2
Gene Aliases:
fi48f03, wu:fi48f03, zgc:109885
Chromosome Location:
Chr.19: 43122612 - 43134877 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03441109_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eef1a1l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039985.1 NP_001035074.1 7 - 8 1329 161
GenBank mRNA BC098530.1 - 7 - 8 1329 161
BC128791.1 - 7 - 8 1308 161

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
nucleotide binding
translation elongation factor activity
GTPase activity
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0