This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03440272_m1
See other GPD1C GE Assays ›
Gene Symbol:
gpd1c
Gene Name:
glycerol-3-phosphate dehydrogenase 1c
Gene Aliases:
fc30a07, gpd1, wu:fc30a07, zgc:63859
Chromosome Location:
Chr.19: 317629 - 325595 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03440272_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gpd1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214753.2 NP_999918.1 1 - 2 144 71
GenBank mRNA BC053116.1 - 1 - 2 77 71
DT076842.1 - 1 - 2 91 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycerol-3-phosphate metabolic process
NADH oxidation
glycerol-3-phosphate catabolic process
oxidation-reduction process
glycerol-3-phosphate dehydrogenase [NAD+] activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0