This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03439906_m1
See other NDRG1A GE Assays ›
Gene Symbol:
ndrg1a
Gene Name:
N-myc downstream regulated 1a
Gene Aliases:
cb775, ndrg1, ndrg1a-1, ndrg1a-2, wu:fb60h02, zgc:63944
Chromosome Location:
Chr.19: 34959967 - 34979837 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03439906_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ndrg1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128353.1 NP_001121825.1 11 - 12 901 104
NM_213348.3 NP_998513.2 13 - 14 953 104
GenBank mRNA AJ579376.1 - 13 - 14 963 104
BC057420.1 - 13 - 14 940 104
LC093846.1 - 11 - 12 875 104
LC093847.1 - 11 - 12 875 104
LC093848.1 - 11 - 12 875 104
LC093849.1 - 11 - 12 875 104
LC093850.1 - 11 - 12 875 104
LC093851.1 - 11 - 12 875 104
LC093852.1 - 13 - 14 937 104
LC093853.1 - 13 - 14 937 104
LC093854.1 - 13 - 14 937 104
LC093855.1 - 13 - 14 937 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s)

signal transduction
peripheral nervous system myelin maintenance
regulation of cell proliferation
mast cell activation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0