This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03439123_g1
See other ALDH3A2A GE Assays ›
Gene Symbol:
aldh3a2a
Gene Name:
aldehyde dehydrogenase 3 family, member A2a
Gene Aliases:
Aldh-III, aldh3a2, wu:fc06b11, zgc:92064
Chromosome Location:
Chr.15: 20830664 - 20839766 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03439123_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: aldh3a2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212649.1 NP_997814.1 5 - 6 754 119
XM_009291620.3 XP_009289895.1 5 - 6 816 119
XM_017359216.2 XP_017214705.1 5 - 6 772 119
GenBank mRNA AF254954.1 - 5 - 6 754 119
BC075877.1 - 5 - 6 743 119
BC155171.1 - 5 - 6 772 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

cellular aldehyde metabolic process
oxidation-reduction process
3-chloroallyl aldehyde dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0