This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03439025_g1
See other RBB4L GE Assays ›
Gene Symbol:
rbb4l
Gene Name:
retinoblastoma binding protein 4, like
Gene Aliases:
Caf1, rbbp7, wu:fa13g08, wu:fb50h10, wu:fc29d09, zgc:56477, zgc:85617
Chromosome Location:
Chr.11: 462621 - 477437 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03439025_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rbb4l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212610.1 NP_997775.1 12 - 13 1330 88
XM_009305881.3 XP_009304156.1 11 - 12 1581 88
GenBank mRNA BC052110.1 - 12 - 13 1330 88
BC067546.1 - 12 - 13 1304 88
BC154778.1 - 12 - 13 1346 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s)

hematopoietic progenitor cell differentiation
DNA replication
chromatin organization
fin regeneration

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0