This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03438901_m1
See other TTN.1 GE Assays ›
Gene Symbol:
ttn.1
Gene Name:
titin, tandem duplicate 1
Gene Aliases:
TTN, hm:zeh1256r, im:6911926, im:7146708, si:dZ167C3.1, si:dz249n21.1, ttnb, ttnl, wu:fc09a12
Chromosome Location:
Chr.9: 42732992 - 42873861 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03438901_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ttn.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021479071.1 XP_021334746.1 92 - 93 21345 67
XM_021479072.1 XP_021334747.1 50 - 51 13744 67
GenBank mRNA LC168152.1 - 82 - 83 21649 67
LC168154.1 - 84 - 85 17758 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
skeletal muscle tissue development
skeletal myofibril assembly
myofibril assembly
sarcomerogenesis
striated muscle myosin thick filament assembly
protein kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0