This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03437572_g1
See other MATR3L1.2 GE Assays ›
Gene Symbol:
matr3l1.2
Gene Name:
matrin 3-like 1.2
Gene Aliases:
MATR3, matr3l, si:bz1d10.1, si:bz1d10.2, wu:fb78d04, wu:fc09f04, wu:fe36a04
Chromosome Location:
Chr.14: 25079782 - 25100000 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03437572_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: matr3l1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122784.2 NP_001116256.2 13 - 14 2528 83
XM_005173122.4 XP_005173179.1 13 - 14 2538 83
XM_005173123.4 XP_005173180.1 13 - 14 2538 83
XM_009291035.3 XP_009289310.1 12 - 13 1637 83
GenBank mRNA CK687062.1 - 3 - 4 201 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0