This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03436615_m1
See other IGF3 GE Assays ›
Gene Symbol:
igf3
Gene Name:
insulin-like growth factor 3
Gene Aliases:
IGF-3
Chromosome Location:
Chr.8: 13978555 - 13985032 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03436615_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: igf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001115050.1 NP_001108522.1 1 - 2 105 71
GenBank mRNA EU272146.1 - 1 - 2 105 71
HQ241070.1 - 1 - 2 105 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
sperm capacitation
insulin-like growth factor receptor binding
hormone activity
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0