This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03436529_m1
See other ARHGAP17B GE Assays ›
Gene Symbol:
arhgap17b
Gene Name:
Rho GTPase activating protein 17b
Gene Aliases:
zgc:172194
Chromosome Location:
Chr.3: 18579800 - 18651305 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03436529_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arhgap17b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114906.2 NP_001108378.2 8 - 9 809 78
NM_001329867.1 NP_001316796.1 8 - 9 895 78
XM_009299322.3 XP_009297597.1 8 - 9 900 78
XM_009299323.3 XP_009297598.1 8 - 9 901 78
XM_009299324.3 XP_009297599.1 8 - 9 901 78
XM_009299325.3 XP_009297600.1 8 - 9 900 78
XM_009299326.3 XP_009297601.1 8 - 9 900 78
XM_021469082.1 XP_021324757.1 8 - 9 901 78
GenBank mRNA BC154580.1 - 8 - 9 809 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
signal transduction
calcium ion regulated exocytosis
regulation of actin cytoskeleton organization
regulation of Rac protein signal transduction
GTPase activator activity
Rac GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0